Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FEZF1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524558

Catalog No. PIPA524558

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Transcription repressor. Involved in the axonal projection and proper termination of olfactory sensory neurons (OSN). Plays a role in rostro-caudal patterning of the diencephalon and in prethalamic formation. Expression is required in OSN to cell-autonomously regulate OSN axon projections. Regulates non-cell-autonomously the layer formation of the olfactory bulb development and the interneurons. May be required for correct rostral migration of the interneuron progenitors.
Specifications

Specifications

FEZF1
Polyclonal
PBS with 0.09% sodium azide
A0PJY2, Q0VDQ9
FEZ, ZNF312B, fez family zinc finger protein 1, zinc finger protein 312B
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
389549, 73191
Western Blot
Unconjugated
FEZF1
Liquid
FEZF1
KLH conjugated synthetic peptide between 67-96 amino acids from the N-terminal region of human FEZF1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit