Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGF-19 Mouse anti-Human, Clone: 117601, R&D Systems™

Mouse Monoclonal Antibody

$119.60 - $839.00

Specifications

Antigen FGF-19
Immunogen E. coli-derived recombinant human FGF-19 Phe27-Lys216 Accession # O95750
Clone 117601
Applications Western Blot, Immunohistochemistry
Conjugate Unconjugated
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB969SP
View Documents
R&D Systems
MAB969SP
25 ug
Each for $119.60
Add to cart
 
MAB969100
View Documents
R&D Systems
MAB969100
100 ug
Each for $356.00
Add to cart
 
MAB969500
View Documents
R&D Systems
MAB969500
500 ug
Each for $839.00
Add to cart
 
Description

Description

FGF-19 Monoclonal antibody MAB969 Specifically detects FGF-19 in Human samples. It is validated for Immunohistochemistry, Western Blotting
Specifications

Specifications

FGF-19
117601
Unconjugated
O95750
Monoclonal
Primary
Detects human FGF-19 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) FGF acidic, rhFGF basic, rhFGF-4, -5, -6, -7, -9, -10, -17, -18, recombinant mouse FGF-8b, -8c, or -15 is observed.
Protein A or G purified from hybridoma culture supernatant
E. coli-derived recombinant human FGF-19 Phe27-Lys216 Accession # O95750
Western Blot, Immunohistochemistry
Purified
Mouse
IgG1
9965
RUO
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit