Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGF R2 (IIIc) Rat anti-Mouse, Clone: 133706, R&D Systems™

Rat Monoclonal Antibody

$114.50 - $839.00

Specifications

Antigen FGFR2
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse FGF R2 alpha (IIIc) Extracellular domains
Clone 133706
Applications Western Blot, Neutralization
Conjugate Unconjugated
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB716SP
View Documents
R&D Systems
MAB716SP
25 ug
Each for $114.50
Add to cart
 
MAB716100
View Documents
R&D Systems
MAB716100
100 ug
Each for $377.00
Add to cart
 
MAB716500
View Documents
R&D Systems
MAB716500
500 ug
Each for $839.00
Add to cart
 
Description

Description

FGF R2 (IIIc) Monoclonal antibody MAB716 Specifically detects FGF R2 (IIIc) in Mouse samples. It is validated for Neutralization, Western Blotting
Specifications

Specifications

FGFR2
133706
Unconjugated
Rat
IgG2a
2263
RUO
Mouse myeloma cell line NS0-derived recombinant mouse FGF R2 alpha (IIIc) Extracellular domains
Western Blot, Neutralization
Purified
Monoclonal
Primary
Detects IIIc isoforms of mouse FGF R2 in direct ELISAs and Western blots. Does not react with IIIb isoforms of recombinant mouse (rm) FGF R2. In direct ELISAs and Western blots, 100% cross-reactivity with IIIc isoforms of recombinant human (rh) FGF R2 is observed, and no cross-reactivity with IIIb isoforms of rhFGF R2 or any isoforms of rhFGF R1, rmFGF R3, or rhFGF R4 is observed.
Protein A or G purified from hybridoma culture supernatant
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit