Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGF10 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA527300
Description
Recommended positive controls: synovial fibroblast, HUVEC, Rat2. Predicted reactivity: Mouse (95%), Rat (100%), Xenopus laevis (83%), Chicken (89%), Sheep (99%), Rhesus Monkey (100%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
FGF10 Polyclonal antibody specifically detects FGF10 in Human, Rat samples. It is validated for Western Blot.Specifications
FGF10 | |
Polyclonal | |
Unconjugated | |
FGF10 | |
AEY17; BB213776; FGF; Fgf10; FGF-10; Fibroblast growth factor; Fibroblast growth factor 10; Gsfaey17; Keratinocyte growth factor 2; produced by fibroblasts of urinary bladder lamina propria | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2255, 25443 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.31 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
O15520, P70492 | |
FGF10 | |
Recombinant protein fragment corresponding to a region within amino acids 18 and 169 of Human FGF10 | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction