Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGF12 Rabbit anti-Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527301

Catalog No. PIPA527301


Add to cart

Description

Description

PA5-27301 targets FGF12 in WB applications and shows reactivity with mouse samples. The PA5-27301 immunogen is recombit protein fragment corresponding to a region within amino acids 1 and 168 of Human FGF12. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth, and invasion. This growth factor lacks the N-terminal signal sequence present in most of the FGF family members, but it contains clusters of basic residues that have been demonstrated to act as a nuclear localization signal. When transfected into mammalian cells, this protein accumulated in the nucleus, but was not secreted. The specific function of this gene has not yet been determined. Two alternatively spliced transcript variants encoding distinct isoforms have been reported.
Specifications

Specifications

FGF12
Polyclonal
Unconjugated
Fgf12
Liquid
FGF12
Recombit protein fragment corresponding to a region within amino acids 1 and 168 of Human FGF12
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Mouse
Western Blot
1.01 mg/mL
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
P61329
FGF12B, FHF1, FGF-12, FHF-1, fibroblast growth factor 12B, fibroblast growth factor FGF-12b, fibroblast growth factor homologous factor 1, myocyte-activating factor
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
14167
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit