Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGF20 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523480

Catalog No. PIPA523480


Add to cart

Description

Description

Fibroblast growth factor 20 belongs to the heparin-binding growth factors family. FGF20 is expressed in normal brain, particularly the cerebellum and in some cancer cell lines. FGF20 has been found to be a critical element in oncogenesis induced by the Wnt signaling pathway. Recombit FGF20 protein induces DNA synthesis in a variety of cell types and is recognized by multiple FGF receptors. Several reports show FGF20 is responsible for Parkinson’s disease.
Specifications

Specifications

FGF20
Polyclonal
Unconjugated
FGF20
Liquid
FGF20
Synthetic peptide (120 GMNDKGELYGSEKLTSEC 137) of FGF20 protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
Q9NP95
FGF-20
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
26281
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit