Learn More
MilliporeSigma™ anti-FGF21 Clone: 2F11, Available on GSA/VA Contract for Federal Government customers only.
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma™ ST1615100UG
Catalog No. ST1615100UG
Add to cart
Description
Description
Specifically detects FGF21 Clone: 2F11 in Human samples, and it is validated for ELISA, Immunohistochemistry (Paraffin), Immuno Blotting
The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities and are involved in a variety of biological processes including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. The function of this growth factor has not yet been determined.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
FGF21
2F11
In PBS, pH 7.2.
Murine
IgG1
RUO
Primary
Recgonizes the FGF21 protein in human tonsil tissue.
ELISA, Immuno Blotting, Immunoblot, Immunohistochemistry (Paraffin)
Unconjugated
Purified
Full-length, recombinant human FGF21 (aa 30-210), expressed as a GST fusion protein
100μg
−20°C
Monoclonal
Human