Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

fibroblast growth factor 21, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human FGF21 protein.

Manufacturer:  Abnova Corporation H00026291B01

Catalog No. 89-007-395


Add to cart

Description

Description

The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities and are involved in a variety of biological processes including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. The function of this growth factor has not yet been determined. [provided by RefSeq

Sequence: MDSDETGFEHSGLWVSVLAGLLLGACQAHPIPDSSPLLQFGGQVRQRYLYTDDAQQTEAHLEIREDGTVGGAADQSPESLLQLKALKPGVIQILGVKTSRFLCQRPDGALYGSLHFDPEACSFRELLLEDGYNVYQSEAHGLPLHLPGNKSPHRDPAPRGPARFLPLPGLPPAPPEPPGILAPQPPDVGSSDPLSMVGPSQGRSPSYAS
Specifications

Specifications

fibroblast growth factor 21
Unconjugated
No additive
BC018404
FGF21
FGF21 (AAH18404.1, 1 a.a. ~ 209 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
26291
Western Blot
Mouse polyclonal antibody raised against a full-length human FGF21 protein.
FGF21
Serum
Mouse
50 uL
Signal Transduction
Y
Polyclonal
Human, Rat
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit