Learn More
fibroblast growth factor 23, Mouse, Polyclonal Antibody, Abnova
Mouse polyclonal antibody raised against a full-length human FGF23 protein.
Manufacturer: Abnova Corporation H00008074B01
Description
The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities and are involved in a variety of biological processes including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. The product of this gene inhibits renal tubular phosphate transport. This gene was identified by its mutations associated with autosomal dominant hypophosphatemic rickets (ADHR), an inherited phosphate wasting disorder. Abnormally high level expression of this gene was found in oncogenic hypophosphatemic osteomalacia (OHO), a phenotypically similar disease caused by abnormal phosphate metabolism. Mutations in this gene have also been shown to cause familial tumoral calcinosis with hyperphosphatemia. [provided by RefSeq
Sequence: MLGARLRLWVCALCSVCSMSVLRAYPNASPLLGSSWGGLIHLYTATARNSYHLQIHKNGHVDGAPHQTIYSALMIRSEDAGFVVITGVMSRRYLCMDFRGNIFGSHYFDPENCRFQHQTLENGYDVYHSPQYHFLVSLGRAKRAFLPGMNPPPYSQFLSRRNEIPLIHFNTPIPRRHTRSAEDDSERDPLNVLKPRARMTPAPASCSQELPSAEDNSPMASDPLGVVRGGRVNTHAGGTGPEGCRPFAKFISpecifications
fibroblast growth factor 23 | |
Unconjugated | |
No additive | |
NM_020638 | |
ADHR|HPDR2|HYPF|PHPTC | |
Mouse | |
50 uL | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Primary | |
8074 |
Western Blot | |
Mouse polyclonal antibody raised against a full-length human FGF23 protein. | |
FGF23 | |
Serum | |
FGF23 | |
FGF23 (NP_065689.1, 1 a.a. ~ 251 a.a) full-length human protein. | |
RUO | |
Y | |
Polyclonal | |
Human |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.