Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KGF Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523702

Catalog No. PIPA523702


Add to cart

Description

Description

This antibody is predicted to react with mouse and rat based on sequence homology.

The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. This protein is a potent epithelial cell-specific growth factor, whose mitogenic activity is predomitly exhibited in keratinocytes but not in fibroblasts and endothelial cells. Studies of mouse and rat homologs of this gene implicated roles in morphogenesis of epithelium, reepithelialization of wounds, hair development and early lung organogenesis.
Specifications

Specifications

KGF
Polyclonal
PBS with 0.09% sodium azide
P21781
HBGF-7, KGF, FGF-7, heparin-binding growth factor 7, keratinocyte growth factor
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
2252
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
FGF7
Liquid
FGF7
KLH conjugated synthetic peptide between 57-86 amino acids from the central region of human FGF7
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit