Learn More
FGFBP3 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA525539
Catalog No. PIPA525539
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
FGFBP3
Polyclonal
Unconjugated
FGFBP3
Liquid
FGFBP3
KLH conjugated synthetic peptide between 181-210 amino acids from the C-terminal region of human FGFBP3
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q8TAT2, Q1HCM0
C10orf13, FGF-BP3, FGF-binding protein 3, FGFBP-3, fibroblast growth factor-binding protein 3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
143282, 72514