Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGFR3 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA514655
Description
FGFR3 is a member of the fibroblast growth factor receptor family, where amino acid sequence is highly conserved between members and throughout evolution. FGFR family members differ from one another in their ligand affinities and tissue distribution. A full-length representative protein would consist of an extracellular region, composed of three immunoglobulin-like domains, a single hydrophobic membrane-spanning segment and a cytoplasmic tyrosine kinase domain. The extracellular portion of the protein interacts with fibroblast growth factors, setting in motion a cascade of downstream signals, ultimately influencing mitogenesis and differentiation. This particular family member binds acidic and basic fibroblast growth hormone and plays a role in bone development and maintece. Mutations in this gene lead to craniosynostosis and multiple types of skeletal dysplasia.
Specifications
FGFR3 | |
Polyclonal | |
Unconjugated | |
P22607 | |
FGFR3 | |
KLH conjugated synthetic peptide between 776-806 amino acids from the C-terminal region of human FGFR3 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
FGFR3 | |
Fibroblast Growth Factor Receptor 3, ACH, CEK2, HSFGFR3EX, JTK4, FGFR-3, fibroblast growth factor receptor 3 variant 4, hydroxyaryl-protein kinase, tyrosine kinase JTK4, FGF Receptor 3 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
2261 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction