Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FHIT Mouse, Unlabeled, Clone: 30, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611740

Catalog No. BDB611740


Add to cart

Description

Description

Multiple tumor-derived cell lines have homozygous deletions in a 200-300kb region in chromsome 3p14.2. This region includes the fragile site locus FRA3B, which may contain deletions in tumor suppressor genes. Abnormalities in the expression and structure of a gene, FHIT, that spans the FRA3B locus have been observed in a variety of carcinomas. FHIT (Fragile Histidine Triad) is widely expressed in normal embryonic and adult tissues and is absent in lung, renal, and gastric tumors. Exposure of FHIT heterozygous knock-out mice to carcinogens produces increased tumorigenicity similar to that observed in Muir-Torre familial cancer syndrome. The only known function of FHIT is as a diadenosine 5 ft.,5 ft.-P1, P3 triphosphate (Ap3A) hydrolase. Both in vitro and in vivo, FHIT regulates the level of Ap3A and adenosine (57 ft.) tetraphosphate(5 ft.) nucleoside. However, this hydrolase activity may not be involved in FHIT tumor suppressor activity, since a mutant FHIT that binds but does not hydrolyze Ap3A is as effective as the wild-type in reducing tumorigenicity in mouse cancer models. Thus, FHIT activity may be essential for a unique antiproliferative signaling pathway.

Host Species: Mouse
Clone: 30
Isotype: IgG1
Species Reactivity [for Features Main]: Mouse
Immunogen: Mouse FHIT aa. 26-148

Immunofluorescence, Western Blotting

Specifications

Specifications

FHIT
30
Unconjugated
Murine
IgG1
RUO
Primary
Murine, Rat
Immunofluorescence, Western Blot
250μg/mL
Affinity Purified
Mouse FHIT aa. 26-148
50μg
Cell Biology
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit