Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FHIT, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA137690
Description
Alterations and deletions of the FHIT gene are strongly linked to the genesis and establishment of human tumors of the lung, cervix, breast, colon, stomach and pancreas. In normal cells, FHIT may act as a tumor suppressor.
Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is normal breast tissue.
Specifications
| FHIT | |
| Polyclonal | |
| FHIT | |
| AP3Aase, FRA3B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2272 | |
| Store at 4°C, do not freeze | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| P49789 | |
| FHIT | |
| Recombinant full length human FHIT protein | |
| 1 mL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction