Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Please call 1-800-766-7000 to place your order or try our site again later.
four and a half LIM domains 2, Mouse, Polyclonal Antibody, Abnova
Mouse polyclonal antibody raised against a full-length human FHL2 protein.
Manufacturer: Abnova Corporation H00002274B01
Description
LIM proteins contain a highly conserved double zinc finger motif called the LIM domain.[supplied by OMIM
Sequence: MTERFDCHHCNESLFGKKYILREESPYCVVCFETLFANTCEECGKPIGCDCKDLSYKDRHWHEACFHCSQCRNSLVDKPFAAKEDQLLCTDCYSNEYSSKCQECKKTIMPGTRKMEYKGSSWHETCFICHRCQQPIGTKSFIPKDNQNFCVPCYEKQHAMQCVQCKKPITTGGVTYREQPWHKECFVCTACRKQLSGQRFTARDDFAYCLNCFCDLYAKKCAGCTNPISGLGGTKYISFEERQWHNDCFNCKKCSLSLVGRGFLTERDDILCPDCGKDISpecifications
four and a half LIM domains 2 | |
Unconjugated | |
No additive | |
NM_201555.1 | |
AAG11|DRAL|SLIM3 | |
Mouse | |
50 uL | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Primary | |
2274 |
Western Blot | |
Mouse polyclonal antibody raised against a full-length human FHL2 protein. | |
FHL2 | |
Serum | |
FHL2 | |
FHL2 (NP_963849.1, 1 a.a. ~ 279 a.a) full-length human protein. | |
RUO | |
Y | |
Polyclonal | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title