Learn More
Fibrillarin Mouse anti-Drosophila, Human, Rat, Clone: 38F3, Invitrogen™ Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen 480009
Catalog No. 48-000-9
Add to cart
Description
Description
This gene product is a component of a nucleolar small nuclear ribonucleoprotein (snRNP) particle thought to participate in the first step in processing preribosomal RNA. It is associated with the U3, U8, and U13 small nuclear RNAs and is located in the dense fibrillar component (DFC) of the nucleolus. The encoded protein contains an N-terminal repetitive domain that is rich in glycine and arginine residues, like fibrillarins in other species. Its central region resembles an RNA-binding domain and contains an RNP consensus sequence. Antisera from approximately 8% of humans with the autoimmune disease scleroderma recognize fibrillarin.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Fibrillarin
Monoclonal
Unconjugated
FBL
Liquid
FBL
Yeast nuclear preparations.
IgG fraction
RUO
Antibody
Monoclonal
Drosophila, Human, Rat
Immunocytochemistry, Immunofluorescence, Western Blot
38F3
Antibody with 10mM sodium azide
P22087, P22509
FIB, FLRN, RNU3IP1, 34 kDa nucleolar scleroderma antigen, 34-kD nucleolar scleroderma antigen, RNA, U3 small nucleolar interacting protein 1, rRNA 2′-O-methyltransferase fibrillarin
Mouse
IgG1
100 μL
Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Primary
2091, 292747, 46234