Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGFR2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA124763
Description
This antibody shows no cross reactivity with FGFR1 or FGFR3. PA1-24763 is expected to cross react with mouse (92% conserved) and rat (92% conserved) due to sequence homology.
FGFR2 Polyclonal antibody specifically detects FGFR2 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
FGFR2 | |
Polyclonal | |
Unconjugated | |
FGFR2 | |
AU043015; AW556123; bacteria-expressed kinase; BBDS; BEK; BEK fibroblast growth factor receptor; BFR-1; CD332; CEK3; CFD1; ECT1; FGF Receptor 2; FGF-10; FGFR2; FGFR-2; Fgfr7; Fgfr-7; fibroblast growth factor receptor 2; JWS; Keratinocyte growth factor receptor; KGFR; KGFRTr; KSAM; K-SAM; protein tyrosine kinase, receptor like 14; Soluble KGFR; svs; TK14; TK25 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
14183, 2263 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1.3 mg/mL | |
0.01M PBS with 1% BSA and 15mM sodium azide; pH 7.4 | |
P21802, P21803 | |
FGFR2 | |
Synthetic peptide, conjugated to KLH by a Glutaraldehyde linker, corresponding to residues K(809) L P Q Y P H I G S V K T(821) of the cytoplasmic region of Human FGFR2 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction