Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGFR3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA137687

Catalog No. 01-673-454


Add to cart

Description

Description

Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is breast or breast carcinoma.

Fibroblast growth factors (FGFs) are members of a large family of structurally related polypeptides that are potent physiological regulators of growth and differentiation for a wide variet of cells of mesodermal, ectodermal and endodermal origin. Four genes encoding for high affinity cell surface FGF receptors (FGFRs) have been identified: FGFR-1, FGFR-2, FGFR-3 and FGFR-4.FGFRs are emembers of the tyrosine kinase family of growth factor receptors. FGFR-3 is widely expressed in many fetal and adult human and animal tissues.
Specifications

Specifications

FGFR3
Polyclonal
PBS with 1% BSA and <0.1% sodium azide
P22607
Fibroblast Growth Factor Receptor 3, ACH, CEK2, HSFGFR3EX, JTK4, FGFR-3, fibroblast growth factor receptor 3 variant 4, hydroxyaryl-protein kinase, tyrosine kinase JTK4, FGF Receptor 3
Rabbit
IgG
1 mL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
2261
Immunohistochemistry (Paraffin)
Unconjugated
FGFR3
Liquid
FGFR3
Synthetic peptide derived from internal region of human FGFR-3 protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit