Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fibronectin Rabbit anti-Human, Mouse, Rat, Porcine, Bovine, Canine, Equine, Feline, Biotin, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP191258B

Catalog No. NBP191258B


Add to cart

Description

Description

Fibronectin Polyclonal antibody specifically detects Fibronectin in Human, Mouse, Rat, Porcine, Bovine, Canine, Equine, Feline samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
Specifications

Specifications

Fibronectin
Biotin
PBS with 0.05% Sodium Azide
Affinity Purified
FN1
A synthetic peptide made toward the C-terminal region of the human Fibronectin protein (within residues 2250-2300). [Swiss-Prot: P02751]
Immunogen affinity purified
RUO
Store at 4C in the dark.
Polyclonal
Human and Mouse. Based on immunogen's sequence homology, this antibody is predicted to react with Chicken (91%), Rat (95%), Canine/Dog (100%), Bovine (100%) and Equine/Horse (100%). Immunogen similarity with other species: Xenopus (86%) and Newt (82%).
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
P02751
CIG, ED-B, fibronectin 1, FINC, FN, FN1, FNZ, GFND, GFND2, LETS, MSF, SMDCF
Rabbit
IgG
0.1 ml
Cancer, Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers
Primary
2335
Bovine, Canine, Equine, Feline, Human, Mouse, Porcine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only