Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fibronectin Mouse anti-Human, Clone: 16E5, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific LFMA0151
Description
A suggested positive control for this product is human plasma.
Fibronectin is a disulfide-bonded dimer with a molecular weight range of 230-250 kDa. In the extracellular matrix of several connective tissues and vessels, fibronectin is present as an insoluble protein that is extensively cross-linked by interchain disulfide bonds forming high molecular mass polymers. Fibronectin is most abundant during embryonic development and tissue remodeling. Fibronectin is also present at high concentrations as a soluble plasma protein. Fibronectin is organized as a linear series of repeating modules which form domains for interaction with fibronectin itself, other matrix components (e.g. collagen and heparin) and receptors on cells (e.g. integrins). Fibronectin is present in a soluble dimeric form in plasma, and in a dimeric or multimeric form at the cell surface and in extracellular matrix. Fibronectin is involved in cell adhesion and migration processes including embryogenesis, wound healing, blood coagulation, host defense, and metastasis. Fibronectin has been implicated in carcinoma development in lung cancer. Further, Fibronectin expression is increased especially in non-small cell lung carcinoma. The adhesion of lung carcinoma cells to Fibronectin enhances tumorgenecity and confers resistance to apoptosis induced by standard chemotherapeutic agents. The gene encoding Fibronectin has three regions subject to alternative splicing, with the potential to produce 20 different transcript variants. However, the full-length nature of some Fibronectin variants has not been determined.Specifications
Fibronectin | |
Monoclonal | |
1 mg/mL | |
FN1 | |
CIG, DKFZp686F10164, DKFZp686H0342, DKFZp686I1370, DKFZp686O13149, ED-B, FINC, FN, FNZ, GFND, GFND2, LETS, MSF | |
Mouse | |
Protein G | |
RUO | |
2335 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunoprecipitation, Western Blot | |
16E5 | |
Unconjugated | |
P02751 | |
FN1 | |
Protein purified from Human plasma | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title