Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ficolin 3 Mouse anti-Human, Clone: 296134, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA524004

Catalog No. PIMA524004


Add to cart

Description

Description

In direct ELISAs and Western blots, no cross-reactivity with Recombinant human Ficolin-2 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

H-Ficolin (Hakata antigen, Ficolin-3) is an innate immunity pattern recognition molecule found in human serum. H-Ficolin binds to distinct pathogen-associated molecular patterns (PAMP) formed by carbohydrates (GlcNAc, GalNAc and fucose) activating complement through the lectin pathway by means of the associated MASPs ((MBL)-associated serine proteases). H-Ficolin consists of 35 kDa protein chains associated in triplet subunits which are again associated in high molecular weight oligomers by means of disulfide bonds at the N-terminal region.
Specifications

Specifications

Ficolin 3
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
O75636
FCN3, FCNH, HAKA1, Collagen/fibrinogen domain-containing protein 3, Hakata antigen, Collagen/fibrinogen domain-containing lectin 3 p35
Mouse
IgG2B
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
8547
Western Blot
296134
Unconjugated
FCN3
Lyophilized
FCN3
Mouse myeloma cell line NS0-derived recombit human Ficolin-3 Isoform 2 Lys22-Arg288
Protein A/G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit