Learn More
c-fos induced growth factor (vascular endothelial growth factor D), Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human FIGF protein.
Supplier: Abnova Corporation H00002277D01P
Description
The protein encoded by this gene is a member of the platelet-derived growth factor/vascular endothelial growth factor (PDGF/VEGF) family and is active in angiogenesis, lymphangiogenesis, and endothelial cell growth. This secreted protein undergoes a complex proteolytic maturation, generating multiple processed forms which bind and activate VEGFR-2 and VEGFR-3 receptors. This protein is structurally and functionally similar to vascular endothelial growth factor C. [provided by RefSeq
Sequence: MYREWVVVNVFMMLYVQLVQGSSNEHGPVKRSSQSTLERSEQQIRAASSLEELLRITHSEDWKLWRCRLRLKSFTSMDSRSASHRSTRFAATFYDIETLKVIDEEWQRTQCSPRETCVEVASELGKSTNTFFKPPCVNVFRCGGCCNEESLICMNTSTSYISKQLFEISVPLTSVPELVPVKVANHTGCKCLPTAPRHPYSIIRRSIQIPEEDRCSHSKKLCPIDMLWDSNKCKCVLQEENPLAGTEDHSHLQEPALCGPHMMFDEDRCECVCKTPCPKDLIQHPKNCSCFECKESLETCCQKHKLFHPDTCSCEDRCPFHTRPCASGKTACAKHCRFPKEKRAAQGPHSRKNPSpecifications
c-fos induced growth factor (vascular endothelial growth factor D) | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human FIGF protein. | |
FIGF | |
VEGF-D/VEGFD | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
Antibody |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_004469 | |
FIGF | |
FIGF (NP_004460.1, 1 a.a. ∼ 354 a.a) full-length human protein. | |
100 μg | |
Angiogenesis | |
2277 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.