Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FKBP10 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA514273

Catalog No. PIPA514273

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The protein belongs to the FKBP-type peptidyl-prolyl cis/trans isomerase family. It is located in endoplasmic reticulum and acts as molecular chaperones.
Specifications

Specifications

FKBP10
Polyclonal
PBS with 0.09% sodium azide
Q96AY3
FK506-binding protein 10; Peptidyl-prolyl cis-trans isomerase; PPIase; Rotamase; Immunophilin FKBP65; 65 kDa FK506-binding protein; FKBP65; PSEC0056; FKBP65; FKBP10
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
60681
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
FKBP10
Liquid
FKBP10
KLH conjugated synthetic peptide between 552-580 amino acids from the C-terminal region of human FKBP10
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit