Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FKBP14 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA513514

Catalog No. PIPA513514

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with rabbit based on sequence homology.

PPIases accelerate the folding of proteins during protein synthesis.
Specifications

Specifications

FKBP14
Polyclonal
PBS with 0.09% sodium azide
Q9NWM8
FK506-binding protein 14; Peptidyl-prolyl cis-trans isomerase; PPIase; Rotamase; 22 kDa FK506-binding protein; FKBP-2; FKBP22; FKBP14
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
55033
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
FKBP14
Liquid
FKBP14
KLH conjugated synthetic peptide between 37-68 amino acids from the N-terminal region of human FKBP14
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit