Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FKBP4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA522277

Catalog No. PIPA522277


Add to cart

Description

Description

PA5-22277 targets FKBP52 in ICC, IF and WB applications and shows reactivity with Human and mouse samples. The PA5-22277 immunogen is a recombit fragment corresponding to a region within amino acids 1 and 207 of FKBP52 (Uniprot IDNo.Q02790). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

FKBP4
Polyclonal
Unconjugated
FKBP4
Liquid
FKBP4
Recombit fragment corresponding to a region within amino acids 1 and 207 of FKBP52 (Uniprot IDNo.Q02790)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Western Blot
0.99 mg/mL
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Q02790, P30416, Q9QVC8
FKBP51; FKBP59; HBI; Hsp56; PPIase; p52; 51 kDa FK506-binding protein; 59 kDa FK506-binding protein 4; HSP binding immunophilin; T-cell FK506-binding protein; peptidyl-prolyl cis-trans isomerase FKBP4; peptidylprolyl cis-trans isomerase; rotamase
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
14228, 2288, 260321
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit