Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FKBP65 Mouse, Unlabeled, Clone: 25, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610648

Catalog No. BDB610648


Add to cart

Description

Description

FK506, one of multiple potent compounds that block T cell proliferation, is used for immunosuppressive therapy. Several FK506-binding proteins (FKBPs) have been identified in the search for intracellular targets for FK506. These include FKBP12, FKBP13, FKBP51, and FKBP52. The immunosuppressive effects of FK506 and FKBPs result from the inhibition of the calcineurin phosphatase, a well known component of the signaling cascade leading to IL-2 production. FKBP65 is another member of the FKBP protein family and contains the characteristic peptidylprolyl cis-trans-isomerase activity. FKBP65 has been reported to be expressed in the lung, testis, brain, heart, and spleen. Although it has a predicted molecular weight of 65 kDa, FKBP65 has been observed to migrate ranging from 68-72 kDa in SDS electrophoresis gels, perhaps, due to glycosylation and/or phosphorylation.

Specifications

Specifications

FKBP65
25
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Western Blot
250μg/mL
FK506 Binding Protein-65
Affinity Purified
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
50μg
Cell Biology
Primary
Human, Murine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit