Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLF11 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA14136

Catalog No. PA14136


Add to cart

Description

Description

PA1-4136 detects FKLF in human samples. PA1-4136 has been successfully used in Western blot procedures. By Western blot this antibody detects FKLF in human fetal liver tissue and K562 erythroid cell line. The PA1-4136 immunogen is a synthetic peptide corresponding to residues E(26) R K R H D S E R S T(36) of human FKLF.

A novel Kruppel-type zinc finger protein, FKLF, that was cloned from globin-expressing human fetal erythroid cells. FKLF acts as a transcriptional activator of embryonic and fetal beta-globin genes.
Specifications

Specifications

KLF11
Polyclonal
Unconjugated
KLF11
Liquid
KLF11
Synthetic peptide corresponding to residues E(26) R K R H D S E R S T(36) of human FKLF.
Affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
O14901
Transforming growth factor-beta-inducible early growth response protein 2, TGFB-inducible early growth response protein 2, TIEG-2, Krueppel-like facto
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
8462
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit