Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FKTN Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP133383
Description
FKTN Polyclonal specifically detects FKTN in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
FKTN | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
FKTN | |
Recombinant protein encompassing a sequence within the center region of human Fukutin. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Tested in human. Not yet tested in any other species. | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot, Immunocytochemistry, Immunofluorescence | |
1.0mg/mL | |
Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000 | |
CMD1X, EC 2.-, FCMDMGC134945, fukutin, Fukuyama type congenital muscular dystrophy (fukutin), Fukuyama type congenital muscular dystrophy protein, Fukuyama-type congenital muscular dystrophy protein, LGMD2MMGC134944, MDDGA4, MDDGB4, MDDGC4, MGC126857, MGC138243 | |
Rabbit | |
54 kDa | |
100 μL | |
Neuroscience | |
2218 | |
Human | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction