Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FLAD1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526206

Catalog No. PIPA526206

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

FLAD1 encodes the enzyme that catalyzes adenylation of flavin mononucleotide (FMN) to form flavin adenine dinucleotide (FAD) coenzyme.
Specifications

Specifications

FLAD1
Polyclonal
PBS with 0.09% sodium azide
Q8NFF5
FAD1, FADS, RP11-307C12.7, FAD pyrophosphorylase, FAD synthase, FAD synthetase, FAD-synthetase, FMN adenylyltransferase, Fad1, flavin adenine dinucleotide synthetase, homolog, flavin adenine dinucleotide synthase, flavin adenine dinucleotide synthetase
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
80308
Western Blot
Unconjugated
FLAD1
Liquid
FLAD1
KLH conjugated synthetic peptide between 551-580 amino acids from the C-terminal region of human FLAD1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit