Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FLAP Mouse, Unlabeled, Clone: 34, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611156

Catalog No. BDB611156


Add to cart

Description

Description

Gelsolin, an actin binding protein, regulates the actin cytoskeleton. Flightless I (fliI), a member of the gelsolin family, was discovered as a mutation in Drosophila that results in flightlessness and, in some cases, lethality. FliI is required for actin organization during myogenesis and embryogenesis. It contains characteristic gelsolin 6-fold segmental repeats and an N-terminal extension of 16 tandem leucine-rich repeats (LRR), which are involved in protein-protein interactions. The human flightless I (FLI) locus lies in a chromosomal region that is deleted in Smith-Magenis syndrome. The C-terminal region of FLI is 31% identical and 52% similar to human gelsolin. An attempt to detect an interaction between FLI and actin resulted in the discovery of FLI LRR Associated Protein (FLAP). FLAP is rich in α-helices and consists of central and C-terminal segments of dimeric coiled coils that are thought to mediate its interaction with FLI LRR. Therefore, FLAP appears to be part of the membrane cytoskeleton and serves as a binding ligand for LRR. This interaction implicates FLI as a linkage between the cytoskeleton and an unidentified intracellular structure.

Host Species: Mouse
Clone: 34
Isotype: IgG1
Species Reactivity: Mouse
Immunogen: Mouse FLAP aa. 516-622
Formula Weight [Chemical]: 85kDa

Immunofluorescence, Western Blotting

Specifications

Specifications

FLAP
34
Unconjugated
Murine
IgG1
RUO
Primary
Human, Murine, Rat
Western Blot
250μg/mL
Affinity Purified
Mouse FLAP aa. 516-622
50μg
Cell Biology
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit