Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flavin reductase Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA528782

Catalog No. PIPA528782


Add to cart

Description

Description

PA5-28782 targets Flavin reductase in WB applications and shows reactivity with Human samples. The PA5-28782 immunogen is synthetic peptide corresponding to a region within amino acids 143 and 206 of Human Flavin reductase. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

The final step in heme metabolism in mammals is catalyzed by the cytosolic biliverdin reductase enzymes A and B (EC 1.3.1.24).
Specifications

Specifications

Flavin reductase
Polyclonal
Unconjugated
BLVRB
Liquid
BLVRB
Synthetic peptide corresponding to a region within amino acids 143 and 206 of Human Flavin reductase
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
0.4 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
P30043
BVRB, FLR, SDR43U1, BVR-B, FR, GHBP, NADPH-dependent diaphorase, NADPH-flavin reductase, biliverdin-IX beta-reductase, flavin reductase (NADPH), green heme-binding protein, short chain dehydrogenase/reductase family 43U, member 1
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
645
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit