Learn More
FLI1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA137705
Description
This antibody is predicted to react with mouse and rat based on sequence homology. Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is angiosarcoma, Ewing's sarcoma or placenta tissue.
ETS-1 is the prototype member of a family of genes identified on the basis of homology to the v-Ets oncogene isolated from the E26 erythroblastosis virus. This family of genes currently includes ETS-1, ETS-2, ERG-1, ERG-2, ELK, E74, FLI-1, PU.1 and PEA3. Members of the ETS gene family exhibit varied pattern of tissue expression, and share a highly conserved carboxy terminal domain containing a sequence related to the SV40 large T antigen nuclear localization signal sequence.
Specifications
FLI1 | |
Polyclonal | |
FLI1 | |
EWSR2, SIC-1, Friend leukemia integration 1 transcription factor, proto-oncogene Fli-1, transcription factor ERGB | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
14247, 2313 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Q01543, P26323 | |
FLI1 | |
Synthetic peptide derived from C-terminus of human FLI-1. | |
1 mL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.