Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-FLIP, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NB10056306SS

 View more versions of this product

Catalog No. NB10056306T


Add to cart

Description

Description

FLIP Polyclonal antibody specifically detects FLIP in Human, Mouse samples. It is validated for Western Blot.
Specifications

Specifications

FLIP
0.5 mg/mL
Western Blot 3 - 5 ug/ml
O15519
CASHFADD-like antiapoptotic molecule 1, CASP8 and FADD-like apoptosis regulator, CASP8AP1, Caspase homolog, Caspase-eight-related protein, Caspase-like apoptosis regulatory protein, caspase-related inducer of apoptosis, CASPER, CasperFLAME1, Cellular FLICE-like inhibitory protein, c-FLIPR, c-FLIPS, CLARPMACH-related inducer of toxicity, FADD-like apoptotic molecule, FLAME, FLAME-1, I-FLICEMRITc-FLIPFLIPc-FLIPL, Inhibitor of FLICE, Usurpin, usurpin beta
Rabbit
IgG
0.025 mg
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse
Western Blot
Unconjugated
PBS with 0.05% BSA with 0.05% Sodium Azide
Affinity Purified
CFLAR
A peptide corresponding to amino acids 206-222 GRSKEQRLKEQLGAQQE of human I-FLICE was used to immunize rabbits.
Immunogen affinity purified
RUO
Primary
8837
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only