Learn More
anti-FLIP, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP177015
Catalog No. NBP177015
Add to cart
Description
Description
FLIP Polyclonal antibody specifically detects FLIP in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
FLIP
1 mg/mL
Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000
AAC51622
CASHFADD-like antiapoptotic molecule 1, CASP8 and FADD-like apoptosis regulator, CASP8AP1, Caspase homolog, Caspase-eight-related protein, Caspase-like apoptosis regulatory protein, caspase-related inducer of apoptosis, CASPER, CasperFLAME1, Cellular FLICE-like inhibitory protein, c-FLIPR, c-FLIPS, CLARPMACH-related inducer of toxicity, FADD-like apoptotic molecule, FLAME, FLAME-1, I-FLICEMRITc-FLIPFLIPc-FLIPL, Inhibitor of FLICE, Usurpin, usurpin beta
Rabbit
IgG
Ion exchange chromatography
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Antibody recognizes the FLIPa only.
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
PBS with 0.02% Sodium Azide
Ion Exchange Chromatography
CFLAR
Antibody was raised against a peptide corresponding to amino acids near the C-terminus of human FLIPaFLIPl form. The immunogen is located within the last 50 amino acids of FLIP.
55 kDa
0.1 mg
Apoptosis, Cancer, Cell Biology, Signal Transduction, Tumor Suppressors
Primary
8837
Human, Mouse, Rat