Learn More
anti-FLIP, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP177020
Catalog No. NBP177020
Add to cart
Description
Description
FLIP Polyclonal antibody specifically detects FLIP in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
Specifications
FLIP
1 mg/mL
Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:500
NP_997536
CASHFADD-like antiapoptotic molecule 1, CASP8 and FADD-like apoptosis regulator, CASP8AP1, Caspase homolog, Caspase-eight-related protein, Caspase-like apoptosis regulatory protein, caspase-related inducer of apoptosis, CASPER, CasperFLAME1, Cellular FLICE-like inhibitory protein, c-FLIPR, c-FLIPS, CLARPMACH-related inducer of toxicity, FADD-like apoptotic molecule, FLAME, FLAME-1, I-FLICEMRITc-FLIPFLIPc-FLIPL, Inhibitor of FLICE, Usurpin, usurpin beta
Rabbit
IgG
Ion exchange chromatography
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
FLIP has short form (FLIPS) and long form (FLIPL) and antibody recognizes the FLIPL only.
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
PBS with 0.02% Sodium Azide
Ion Exchange Chromatography
CFLAR
Antibody was raised against a 17 amino acid peptide near the carboxy terminus of mouse FLIP. The immunogen is located within the last 50 amino acids of FLIP.
58 kDa
0.1 mg
Apoptosis, Cancer, Cell Biology, Signal Transduction, Tumor Suppressors
Primary
8837
Human, Mouse