Learn More
FLIP Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier:  Thermo Scientific PA130801
Catalog No. PIPA130801
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Description
Description
PA1-30801 detects FLIP from mouse, human, rat samples. PA1-30801 has been successfully used in Western blot applications. The PA1-30801 immunogen is: Peptide corresponding to amino acids 2 to 18 of human FLIP2. The sequence is identical in all FLIP splice variants.
FLIP (CFLAR) is an apoptosis regulator protein which functions as a crucial link between cell survival and cell death pathways in mammalian cells. It acts as an inhibitor of TNFRSF6 mediated apoptosis. A proteolytic fragment (p43) is likely retained in the death-inducing signaling complex (DISC) thereby blocking further recruitment and processing of caspase-8 at the complex. Full length and shorter isoforms have been shown either to induce apoptosis or to reduce TNFRSF-triggered apoptosis. Lacks enzymatic (caspase) activity.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FLIP
Polyclonal
Unconjugated
O15519, O35732
CFLAR
Peptide corresponding to amino acids 2 to 18 of human FLIP2. The sequence is identical in all FLIP splice variants.
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG
Immunocytochemistry, Immunofluorescence, Western Blot
0.5 mg/mL
CFLAR
Casper, Inhibitor of FLICE, I-FLICE, FLAME-1,CASH, CLARP, c-FLIP, MRIT, CASH, Usurpin
Rabbit
IgG fraction
RUO
117279, 12633, 8837
4° C
Liquid
Product Suggestions
Product Suggestions