Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FLIP Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA141242

Catalog No. 01-675-596


Add to cart

Description

Description

Suggested positive control: Hela whole cell extract, HeLa cell lysate.

FLIP (CFLAR) is an apoptosis regulator protein which functions as a crucial link between cell survival and cell death pathways in mammalian cells. It acts as an inhibitor of TNFRSF6 mediated apoptosis. A proteolytic fragment (p43) is likely retained in the death-inducing signaling complex (DISC) thereby blocking further recruitment and processing of caspase-8 at the complex. Full length and shorter isoforms have been shown either to induce apoptosis or to reduce TNFRSF-triggered apoptosis. Lacks enzymatic (caspase) activity.
Specifications

Specifications

FLIP
Polyclonal
Unconjugated
CFLAR
Liquid
CFLAR
Synthetic peptide corresponding to residues G(206) R S K E Q R L K E Q L G A Q Q E(222) of human I-FLICE. 
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
O15519, O35732
I-FLICE
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
12633, 8837
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit