Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
hypothetical protein FLJ10357, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human FLJ10357 protein.
Manufacturer: Abnova Corporation H00055701B01
Description
Sequence: MEAGPYLPRALQQPLEQLTRYGRLLEELLREAGPELSSECRALGAAVQLLREQEARGRDLLAVEAVRGCEIDLKEQGQLLHRDPFTVICGRKKCLRHVFLFEHLLLFSKLKGPEGGSEMFVYKQAFKTADMGLTENIGDSGLCFELWFRRRRAREAYTLQATSPEIKLKWTSSIAQLLWRQAAHNKELRVQQMVSMGIGNKPFLDIKALGERTLSALLTGRAARPRASVAVSSFEHAGPSLPGLSPGACSLPARVEEEAWDLDVKQISLAPETLDSSGDVSPGPRNSPSLQPPHPGSSTPTLASRGILGLSRQSHARALSDPTTPLSpecifications
hypothetical protein FLJ10357 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human FLJ10357 protein. | |
FLJ10357 | |
SOLO | |
Mouse | |
50 μL | |
Yes | |
55701 | |
Serum |
Western Blot | |
Unconjugated | |
No additive | |
BC000084.2 | |
FLJ10357 | |
FLJ10357 (AAH00084.3, 1 a.a. ∼ 326 a.a) full-length human protein. | |
RUO | |
Primary | |
Human |
Safety and Handling
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title