Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-oxoglutarate and iron-dependent oxygenase domain containing 1, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human OGFOD1 protein.

Manufacturer:  Abnova Corporation H00055239B01

Catalog No. 89-007-828


Add to cart

Description

Description

Sequence: MNGKRPAEPGPARVGKKGKKEVMAEFSDAVTEETLKKQVAEAWSRRTPFSHEVIVMDMDPFLHCVIPNFIQSQDFLEGLQKELMNLDFHEKYNDLYKFQQSDDLKKRREPHISTLRKILFEDFRSWLSDISKIDLESTIDMSCAKYEFTDALLCHDDELEGRRIAFILYLVPPWDRSMGGTLDLYSIDEHFQPKQIVKSLIPSWNKLVFFEVSPVSFHQVSEVLSEEKSRLSISGWFHGPSLTRPPNYFEPPIPRSPHIPQDHEILYDWINPTYLDMDYQVQIQEEFEESSEILLKEFLKPEKFTKVCEALEHGHVEWSSRGPPNKRFYEKAEESKLPEILKECMKLFRSEALFLLLSNFTGLKLHFLAPSEEDEMNDKKEAETTDITEEGTSHSPPEPENNQMAISNNSQQSNEQTDPEPEENETKKESSVPMCQGELRHWKTGHYTLIHDHSKAEFALDLILYCGCEGWEPEYGGFTSYIAKGEDEELLTVNPESNSLALVYRDRETLKFVKHINHRSLEQKKTFPNRTGFWDFSFIYYE
Specifications

Specifications

2-oxoglutarate and iron-dependent oxygenase domain containing 1
Unconjugated
No additive
NM_018233.3
FLJ10826|KIAA1612|TPA1
Mouse
50 uL
Endocrine/Metabolism
Y
Polyclonal
Human
Immunofluorescence, Western Blot
Mouse polyclonal antibody raised against a full-length human OGFOD1 protein.
OGFOD1
Serum
OGFOD1
OGFOD1 (NP_060703.3, 1 a.a. ~ 542 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
55239
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit