Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-FLJ11184, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP216505

Catalog No. NBP216505


Add to cart

Description

Description

FLJ11184 Polyclonal specifically detects FLJ11184 in Human samples. It is validated for Western Blot.
Specifications

Specifications

FLJ11184
1mg/mL
Western Blot 1:500-1:3000
Q96EY4
chromosome 4 open reading frame 43, FLJ11184, FLJ52950, hypothetical protein LOC55319
Rabbit
IgG
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Human
Western Blot
Unconjugated
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
Affinity Purified
TMA16
Recombinant protein encompassing a sequence within the center region of human FLJ11184. The exact sequence is proprietary.
24 kDa
0.1 ml
Stem Cell Markers
Primary
55319
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only