Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AAGAB Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525397

Catalog No. PIPA525397

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

FLJ11506 may play a role in membrane traffic.
Specifications

Specifications

AAGAB
Polyclonal
PBS with 0.09% sodium azide
Q6PD74
p34, alpha- and gamma-adaptin-binding protein p34
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
79719
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
AAGAB
Liquid
AAGAB
KLH conjugated synthetic peptide between 286-313 amino acids from the C-terminal region of human FLJ11506
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit