Learn More
anti-Flt-3/Flk-2, PE, Clone: 66903, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB812P
Catalog No. FAB812P
Add to cart
This item is not returnable. View return policy
Description
Description
Applications:
Flow Cytometry
Specifications
Specifications
Flt-3/Flk-2/CD135
66903
Flow Cytometry 10 uL/10^6 cells
AAA18947
CD135, CD135 antigen, EC 2.7.10, fetal liver kinase 2, FL cytokine receptor, Flk-2, FLK2, Flt3, FLT-3, FLT3 receptor tyrosine kinase, Fms-like tyrosine kinase 3, fms-related tyrosine kinase 3, growth factor receptor tyrosine kinase type III, Stem cell tyrosine kinase 1, STK-1, STK1EC 2.7.10.1, Tyrosine-protein kinase receptor FLT3
Mouse
IgG1
100 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detect human Flt-3/Flk-2 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse Flt-3, recombinant human (rh) SCF R, rhPDGF R alpha , or rhPDGF R beta is observed.
Flow Cytometry
R-PE
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified
FLT3
Mouse myeloma cell line NS0-derived recombinant human Flt-3/Flk-2 Asn27-Asn541 Accession # AAA18947
Protein A or G purified from ascites
RUO
Primary
2322
Human