Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FLT3 Rat anti-Mouse, PE, Clone: 113308, Invitrogen™

Rat Monoclonal Antibody

Manufacturer:  Invitrogen MA523648

Catalog No. PIMA523648


Add to cart

Description

Description

In direct ELISAs, no cross-reactivity with recombit human Flt-3 is observed. Endoxin level is <0.10 EU per 1 µg of the antibody by the LAL method.

This gene encodes a class III receptor tyrosine kinase that regulates hematopoiesis. The receptor consists of an extracellular domain composed of five immunoglobulin-like domains, one transmembrane region, and a cytoplasmic kinase domain split into two parts by a kinase-insert domain. The receptor is activated by binding of the fms-related tyrosine kinase 3 ligand to the extracellular domain, which induces homodimer formation in the plasma membrane leading to autophosphorylation of the receptor. The activated receptor kinase subsequently phosphorylates and activates multiple cytoplasmic effector molecules in pathways involved in apoptosis, proliferation, and differentiation of hematopoietic cells in bone marrow. Mutations that result in the constitutive activation of this receptor result in acute myeloid leukemia and acute lymphoblastic leukemia.
Specifications

Specifications

FLT3
Monoclonal
PE
Flt3
Liquid
FLT3
Mouse myeloma cell line NS0-derived recombit mouse Flt-3/Flk-2 Asn28-Ser544 (predicted)
Protein A/G
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
113308
Buffered saline solution with BSA and 0.09% sodium azide
Q00342
FLT3, Fms-like tyrosine kinase 3, receptor-type tyrosine-protein kinase FLT3, CD135, FL cytokine receptor, fetal liver kinase 2, stem cell tyrosine kinase 1
Rat
IgG2A
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
14255
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit