Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VEGF Receptor 3, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA128157

Catalog No. PA128157

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene encodes a tyrosine kinase receptor for vascular endothelial growth factors C and D. The protein is thought to be involved in lymphangiogenesis and maintenance of the lymphatic endothelium. Mutations in this gene cause hereditary lymphedema type IA.

PA1-28157 detects FLT4 from mouse, rat, human samples.PA1-28157 has been successfully used in immunohistochemistry (paraffin tissue) applications.The PA1-28157 immunogen is: Synthetic peptide derived from C-terminus of human Flt4.
Specifications

Specifications

FLT4
Polyclonal
Unconjugated
FLT4
Purified
FLT4
Synthetic peptide derived from C-terminus of human Flt4.
IgG fraction
RUO
Antibody
Polyclonal
Human, Murine, Rat
Immunohistochemistry (Paraffin)
0.1mg/mL
PBS with 0.1% sodium azide; pH 7.4
P35916, P35917, Q91ZT1
FLT41, LMPH1A, PCL, VEGFR3
Rabbit
IgG
250μL
Store at 4°C
Primary
2324
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit