Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FMN2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA523251
Description
FMN2 Polyclonal antibody specifically detects FMN2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
FMN2 | |
Polyclonal | |
Unconjugated | |
FMN2 | |
AU024104; FMN2; formin 2; formin-2; RP11-90L13.1 | |
Rabbit | |
Protein A | |
RUO | |
56776 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
Q9NZ56 | |
FMN2 | |
A synthetic peptide corresponding to amino acids 657-688 of human FMN2 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction