Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

formin-like 1, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human FMNL1 protein.

Manufacturer:  Abnova Corporation H00000752B01

Catalog No. 89-005-009


Add to cart

Description

Description

This gene encodes a formin-related protein. Formin-related proteins have been implicated in morphogenesis, cytokinesis, and cell polarity. An alternative splice variant has been described but its full length sequence has not been determined. [provided by RefSeq

Sequence: MPLLNWVALKPSQITGTVFTELNDEKVLQELDMSDFEEQFKTKSQGPSLDLSALKSKAAQKAPSKATLIEANRAKNLAITLRKGNLGAERICQAIEAYDLQALGLDFLELLMRFLPTEYERSLITRFEREQRPMEELSEEDRFMLCFSRIPRLPERMTTLTFLGNFPDTAQLLMPQLNAIIAASMSIKSSDKLRQILEIVLAFGNYMNSSKRGAAYGFRLQSLDALLEMKSTDRKQTLLHYLVKVIAEKYPQLTGFHSDLHFLDKAGSVSLDSVLADVRSLQRGLELTQREFVRQDDCMVLKEFLRANSPTMDKLLADSKTAQEAFESVVEYFGENPKTTSPGLFFSLFSRFIKAYKKAEQEVEQWKKEAAAQEAGADTPGKGEPPAPKSPPKARRPQMDLISELKRRQQKEPLIYESDRDGAIEDIITDLRNQPYIRADTGRRSARRRPPGPPLQVTSEVSL
Specifications

Specifications

formin-like 1
Unconjugated
No additive
BC021906.1
C17orf1|C17orf1B|FHOD4|FMNL|KW-13|MGC133052|MGC1894|MGC21878
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
752
Western Blot
Mouse polyclonal antibody raised against a full-length human FMNL1 protein.
FMNL1
Serum
FMNL1
FMNL1 (AAH21906.1, 1 a.a. ~ 463 a.a) full-length human protein.
RUO
Y
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit