Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FMNL3 Rabbit anti-Human, Mouse, Rat, Canine, Equine, Primate, Rhesus Macaque, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication

Manufacturer:  Novus Biologicals NBP224724SS

 View more versions of this product

Catalog No. NBP224724SS


Add to cart

Description

Description

FMNL3 Polyclonal specifically detects FMNL3 in Human, Mouse, Rat, Canine, Equine, Primate, Rhesus Macaque samples. It is validated for Western Blot.
Specifications

Specifications

FMNL3
0.5 mg/mL
Western Blot 0.1-0.5 ug/ml
Q8IVF7
formin-like 3
Rabbit
IgG
0.025 mg
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
100% homologous in human (isoforms CRA_a and CRA_f).
Western Blot
Unconjugated
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Purified
FMNL3
A portion of amino acids 150-200 of human Formin-like protein 3 was used as the immunogen.
Protein G purified
RUO
Primary
91010
Human, Mouse, Rat, Canine, Equine, Primate, Rhesus Macaque
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only