Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FMNL3 Rabbit anti-Human, Mouse, Rat, Canine, Equine, Primate, Rhesus Macaque, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$119.60 - $293.00

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acids 150-200 of human Formin-like protein 3 was used as the immunogen.
Antigen FMNL3
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP224724SS
View Documents
Novus Biologicals
NBP224724SS
0.025 mg
Each for $119.60
Add to cart
 
NBP224724
View Documents
Novus Biologicals
NBP224724
0.1 mg
Each for $293.00
Add to cart
 
Description

Description

FMNL3 Polyclonal antibody specifically detects FMNL3 in Human, Mouse, Rat, Canine, Equine, Primate, Rhesus Macaque samples. It is validated for Western Blot.
Specifications

Specifications

Rabbit
A portion of amino acids 150-200 of human Formin-like protein 3 was used as the immunogen.
Polyclonal
Primary
Western Blot
Q8IVF7
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
IgG
FMNL3
Purified
RUO
100% homologous in human (isoforms CRA_a and CRA_f).
91010
0.5 mg/mL
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit