Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-FMO2, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP133318

Catalog No. NBP133318


Add to cart

Description

Description

FMO2 Polyclonal antibody specifically detects FMO2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

FMO2
1.0mg/mL
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Affinity Purified
FMO2
Recombinant protein encompassing a sequence within the center region of human FMO2. The exact sequence is proprietary.
54 kDa
100 ul
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Not yet tested in any other species.
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
dimethylaniline monooxygenase [N-oxide-forming] 2, Dimethylaniline oxidase 2, EC 1.14.13.8, flavin containing monooxygenase 2, flavin containing monooxygenase 2 (non-functional), FLJ40826, FMO 1B1, FMO 2, FMO, pulmonary, FMO1B1, Pulmonary flavin-containing monooxygenase 2
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
2327
Human
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only