Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fibromodulin Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526250

Catalog No. PIPA526250


Add to cart

Description

Description

This antibody is predicted to react with bovine based on sequence homology.

Fibromodulin is a member of a family of small interstitial proteoglycans, containing a central region composed of leucine-rich repeats with 4 keratan sulfate chains flanked by disulfide-bonded terminal domains. It may participate in the assembly of the extracellular matrix as it interacts with type I and type II collagen fibrils and inhibits fibrillogenesis in vitro. It may also regulate TGF-beta activities by sequestering TGF-beta into the extracellular matrix.
Specifications

Specifications

Fibromodulin
Polyclonal
PBS with 0.09% sodium azide
Q06828, P50608
SLRR2E, FM, KSPG fibromodulin, collagen-binding 59 kDa protein, fibromodulin proteoglycan, keratan sulfate proteoglycan fibromodulin
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
14264, 2331
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
FMOD
Liquid
FMOD
KLH conjugated synthetic peptide between 337-364 amino acids from the C-terminal region of human FMOD
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit